4-Ethynylpiperidine hydrochloride 95%

Supplier: Ambeed
Warning

A409340-100MG A409340-1G A409340-250MG A409340-25G A409340-5G
76990-310EA 10 USD
76990-310 76990-312 76990-314 77890-843 77890-844
4-Ethynylpiperidine hydrochloride 95%
4-Ethynylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂ClN
MDL Number: MFCD18910999
CAS Number: 550378-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR