5-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A408746-1G A408746-250MG A408746-5G
76990-300EA 115 USD
76990-300 76990-302 76990-304
5-(Methoxycarbonyl)indole-2-boronic acid pinacol ester 95%
5-(Methoxycarbonyl)indole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₀BNO₄
MDL Number: MFCD11858383
CAS Number: 1256358-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR