2'-O-Methylguanosine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-9-((2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-1H-purin-6(9H)-one

A408549-100MG A408549-1G A408549-1MG A408549-250MG A408549-5G A408549-25G
77004-006EA 21.22 USD
77004-006 77004-008 77004-010 77004-012 77004-014 77672-994
2'-O-Methylguanosine 98%
2'-O-Methylguanosine

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₅O₅
MW: 297.27 g/mol
MDL Number: MFCD00057053
CAS Number: 2140-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR