Ethyl-4-chloro-6-fluoroquinoline-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A408331-5G
A408331-1G
A408331-250MG
77033-724EA
70
USD
77033-724
77872-645
77872-646
Ethyl-4-chloro-6-fluoroquinoline-3-carboxylate 98%
Ethyl 4-chloro-6-fluoroquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₉ClFNO₂ |
MDL Number:
MFCD00173350 CAS Number: 77779-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...