2,6-Difluoro-4-methoxybenzylamine 98%

Supplier: Ambeed
Danger

A408132-1G A408132-100MG A408132-250MG
77016-352EA 64 USD
77016-352 77890-838 77890-839
2,6-Difluoro-4-methoxybenzylamine 98%
2,6-Difluoro-4-methoxybenzylamine

Precaución: for Research Use Only

Formula: C₈H₉F₂NO
MDL Number: MFCD04115924
CAS Number: 771573-20-7
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR