N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide 96%

Supplier: Ambeed
Warning

Synonyms: Desbutylbupivacaine, N-Desbutyl bupivacaine, N-(2,6-Dimethylphenyl)piperidine-2-carboxamide

A407763-100G A407763-10G A407763-25G A407763-5G
77003-994EA 62.6 USD
77003-994 77003-996 77003-998 77004-000
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide 96%
N-(2',6'-Dimethylphenyl)-2-piperidinecarboxamide

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O
MW: 232.32 g/mol
MDL Number: MFCD01701244
CAS Number: 15883-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR