2,4-Dichloro-N-pentylbenzamide 97%

Supplier: Ambeed
Warning

A407736-1G A407736-250MG
77033-720EA 100 USD
77033-720 77033-722
2,4-Dichloro-N-pentylbenzamide 97%
2,4-Dichloro-N-pentylbenzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂NO
MDL Number: MFCD00454449
CAS Number: 2447-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR