(5-Phenyl-1H-pyrazol-3-yl)methanol 96%

Supplier: Ambeed
Warning

A407259-1G A407259-250MG
77003-986EA 102.91 USD
77003-986 77003-988
(5-Phenyl-1H-pyrazol-3-yl)methanol 96%
(5-Phenyl-1H-pyrazol-3-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD11046186
CAS Number: 179057-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR