2,3,4-Trifluorocinnamic acid 95%

Supplier: Ambeed
Warning

A406125-1G A406125-250MG A406125-5G
77003-980EA 54 USD
77003-980 77003-982 77003-984
2,3,4-Trifluorocinnamic acid 95%
2,3,4-Trifluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₂
MDL Number: MFCD00083552
CAS Number: 207742-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR