(4-Bromo-2-ethoxy-6-fluorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

A405627-100MG A405627-1G
77003-976EA 23.34 USD
77003-976 77003-978
(4-Bromo-2-ethoxy-6-fluorophenyl)boronic acid 95%
(4-Bromo-2-ethoxy-6-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BBrFO₃
MW: 262.87 g/mol
MDL Number: MFCD12026730
CAS Number: 1315340-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR