3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine 98%

Supplier: Ambeed
Warning

A405611-100MG A405611-1G A405611-250MG
77016-322EA 31.83 USD
77016-322 77016-324 77016-326
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine 98%
3-Bromo-6-chloro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
MDL Number: MFCD09749982
CAS Number: 1000341-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR