4,7-Dibromo-5,6-di-n-octyloxy-2,1,3-benzothiadiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4,7-Dibromo-5,6-bis(octyloxy)benzo[c][1,2,5]thiadiazole

A405119-100MG A405119-1G A405119-250MG
76990-288EA 28 USD
76990-288 76990-290 76990-292
4,7-Dibromo-5,6-di-n-octyloxy-2,1,3-benzothiadiazole 95%
4,7-Dibromo-5,6-di-n-octyloxy-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₂₂H₃₄Br₂N₂O₂S
MDL Number: MFCD19940667
CAS Number: 1192352-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR