3-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A405004-250MG A405004-5G A405004-1G
76990-284EA 125 USD
76990-284 76990-286 77568-820
3-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
3-Methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Precaución: For research use only.

Formula: C₆H₇N₅
MDL Number: MFCD13813768
CAS Number: 5399-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR