3-Bromo-6-mercaptopyridine 95%

Supplier: Ambeed
Danger

A404988-10G A404988-250MG A404988-5G A404988-1G
77033-698EA 85 USD
77033-698 77033-700 77033-702 77594-228
3-Bromo-6-mercaptopyridine 95%
3-Bromo-6-mercaptopyridine

Precaución: for Research Use Only

Formula: C5H4BrNS
MDL Number: MFCD09729468
CAS Number: 56673-34-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR