Boc-(S)-2-amino-3-ethoxypropionic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-3-ethoxypropanoic acid
A404978-100MG
A404978-1G
A404978-250MG
77003-970EA
46.68
USD
77003-970
77003-972
77003-974
Boc-(S)-2-amino-3-ethoxypropionic acid 95%
Boc-(S)-2-amino-3-ethoxypropionic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₅ MW: 233.27 g/mol |
MDL Number:
MFCD06797558 CAS Number: 104839-00-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...