2-Chloro-1-iodo-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A404842-100G A404842-10G A404842-1G A404842-25G A404842-5G
77003-960EA 150 USD
77003-960 77003-962 77003-964 77003-966 77003-968
2-Chloro-1-iodo-5-nitrobenzene 95%
2-Chloro-1-iodo-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MDL Number: MFCD11521301
CAS Number: 74534-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR