trans-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

A404802-100MG A404802-1G A404802-250MG
76990-270EA 66.99 USD
76990-270 76990-272 76990-274
trans-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%
trans-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD09927457
CAS Number: 136315-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR