4-(Tetrahydropyran-4-yloxy)aniline 95%

Supplier: Ambeed
Warning

A402803-100MG A402803-250MG A402803-1G
77033-678EA 63 USD
77033-678 77033-680 77668-838
4-(Tetrahydropyran-4-yloxy)aniline 95%
4-(Tetrahydropyran-4-yloxy)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD09064968
CAS Number: 917483-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR