4-Bromobutyric acid 98%

Supplier: Ambeed
Danger

A402452-500G A402452-100G
77003-938EA 5.31 USD
77003-938 77594-220 77882-071
4-Bromobutyric acid 98%
4-Bromobutyric acid

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₂COOH
MDL Number: MFCD00002817
CAS Number: 2623-87-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR