N-t-BOC-cis-4-Fluoro-L-prolinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: (2S,4S)-tert-Butyl 2-cyano-4-fluoropyrrolidine-1-carboxylate

A402440-1G A402440-250MG
77003-934EA 50.93 USD
77003-934 77003-936
N-t-BOC-cis-4-Fluoro-L-prolinonitrile 97%
N-t-BOC-cis-4-Fluoro-L-prolinonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₅FN₂O₂
MW: 214.237 g/mol
MDL Number: MFCD09263600
CAS Number: 426844-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR