4-Bromobenz[a]anthracene 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromotetraphene

A402297-100MG A402297-1G A402297-250MG
77016-306EA 15.92 USD
77016-306 77016-308 77016-310
4-Bromobenz[a]anthracene 95%
4-Bromobenz[a]anthracene

Precaución: for Research Use Only

Formula: C₁₈H₁₁Br
MW: 307.19 g/mol
MDL Number: MFCD25371993
CAS Number: 61921-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR