tert-Butyl 3,6-diazabicyclo[3.2.2]nonane-6-carboxylate 95%

Supplier: Ambeed
Warning

A401583-100MG A401583-1G A401583-250MG
77033-666EA 141.1 USD
77033-666 77033-668 77033-670
tert-Butyl 3,6-diazabicyclo[3.2.2]nonane-6-carboxylate 95%
tert-Butyl 3,6-diazabicyclo[3.2.2]nonane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD23099510
CAS Number: 1214743-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR