Pyrazine-2-carboxylic acid 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid
A401180-100G
A401180-10G
A401180-250MG
A401180-25G
A401180-500G
A401180-1000G
77016-296EA
38
USD
77016-296
77016-298
77016-300
77016-302
77016-304
77684-136
Pyrazine-2-carboxylic acid 98%, Ambeed.Inc
Pyrazine-2-carboxylic acid
Precaución: For research use only.
Formula:
C₅H₄N₂O₂ MW: 124.1 g/mol Boiling Pt: 313 |
MDL Number:
MFCD00006130 CAS Number: 98-97-5 Merck Index: 12,08143 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...