[3-(Ethoxycarbonyl)prop-1-yl]tris(phenyl)phosphonium bromide 95%

Supplier: Ambeed
Warning

Synonyms: [4-Ethoxy-4-oxo(but-1-yl)]tris(phenyl)phosphonium bromide, (4-Ethoxy-4-oxobutyl)triphenylphosphonium bromide

A401165-100G A401165-25G
77033-658EA 229.16 USD
77033-658 77033-660
[3-(Ethoxycarbonyl)prop-1-yl]tris(phenyl)phosphonium bromide 95%
[3-(Ethoxycarbonyl)prop-1-yl]tris(phenyl)phosphonium bromide

Precaución: for Research Use Only

Formula: C₂₄H₂₆O₂P·Br
MW: 457.34 g/mol
MDL Number: MFCD00274610
CAS Number: 50479-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR