Ethyl 4-bromo-2-chlorobenzoate 98%

Supplier: Ambeed
Warning

A400852-1G A400852-250MG A400852-5G
77016-290EA 133.68 USD
77016-290 77016-292 77016-294
Ethyl 4-bromo-2-chlorobenzoate 98%
Ethyl 4-bromo-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.51 g/mol
MDL Number: MFCD10000926
CAS Number: 76008-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR