2-Nitro-5-phenoxyaniline 98%

Supplier: Ambeed
Warning

A400744-1G A400744-250MG
77016-286EA 38.2 USD
77016-286 77016-288
2-Nitro-5-phenoxyaniline 98%
2-Nitro-5-phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₃
MW: 230.22 g/mol
MDL Number: MFCD03788969
CAS Number: 1552-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR