2-Methyl-4-pyrimidinamine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpyrimidin-4-amine

A400601-10G A400601-1G A400601-5G
77033-644EA 431.79 USD
77033-644 77033-646 77033-648
2-Methyl-4-pyrimidinamine 97%
2-Methyl-4-pyrimidinamine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.1292 g/mol
MDL Number: MFCD00626295
CAS Number: 74-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR