(1S,2S)-(+)-1-Amino-2-indanol 97%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-1-Amino-2-indanol, (1S,2S)-(+)-1-Amino-2-hydroxyindan

A400496-1G A400496-250MG A400496-5G
77003-908EA 51.99 USD
77003-908 77003-910 77003-912
(1S,2S)-(+)-1-Amino-2-indanol 97%
(1S,2S)-(+)-1-Amino-2-indanol

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD08275383
CAS Number: 163061-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR