4-Ethynyl-2-methylthiazole 97%

Supplier: Ambeed
Warning

A400284-100MG A400284-250MG A400284-1G
76990-244EA 75 USD
76990-244 76990-246 77890-824
4-Ethynyl-2-methylthiazole 97%
4-Ethynyl-2-methylthiazole

Precaución: for Research Use Only

Formula: C₆H₅NS
MDL Number: MFCD18207414
CAS Number: 107263-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR