4'-Methylacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: Methyl p-tolyl ketone, p-Methylacetophenone, 1-(p-Tolyl)ethanone

A400211-1000G A400211-100G A400211-25G A400211-500G
77003-900EA 70.02 USD
77003-900 77003-902 77003-904 77003-906
4'-Methylacetophenone 97%
4'-Methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00008751
CAS Number: 122-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR