4-Bromo-2,3-xylidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-2,3-dimethylaniline
A400170-10G
A400170-1G
A400170-250MG
A400170-25G
A400170-5G
77033-634EA
123.07
USD
77033-634
77033-636
77033-638
77033-640
77033-642
4-Bromo-2,3-xylidine 98%
4-Bromo-2,3-xylidine
Precaución: for Research Use Only
Formula:
C₈H₁₀BrN MW: 200.0800 g/mol |
MDL Number:
MFCD08689504 CAS Number: 22364-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...