Pyrimidine-4,6-dicarbonitrile 95%

Supplier: Ambeed
Warning

A399788-100MG A399788-250MG
76990-224EA 23.34 USD
76990-224 76990-226
Pyrimidine-4,6-dicarbonitrile 95%
Pyrimidine-4,6-dicarbonitrile

Precaución: for Research Use Only

Formula: C₆H₂N₄
MW: 130.11 g/mol
MDL Number: MFCD11870748
CAS Number: 50844-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR