2-Amino-5-(diethylamino)toluene monohydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Diethylamino)-o-toluidine Monohydrochloride, 2-Amino-5-(diethylamino)toluene Monohydrochloride, 2-Methyl-4-(diethylamino)aniline Monohydrochloride 4-(Diethylamino)-o-toluidine Monohydrochloride, 2-Amino-5-(diethylamino)toluene hydrochloride, 2-Methyl-4-(diethylamino)aniline Monohydrochloride

A399512-25G A399512-5G
77003-890EA 23.34 USD
77003-890 77003-892
2-Amino-5-(diethylamino)toluene monohydrochloride 98%
2-Amino-5-(diethylamino)toluene monohydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂
MW: 214.74 g/mol
MDL Number: MFCD00035466
CAS Number: 2051-79-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR