1-(4-Bromophenyl)ethylamine 98%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine

A399489-10G A399489-1G A399489-5G A399489-25G
77003-884EA 26.53 USD
77003-884 77003-886 77003-888 77601-760
1-(4-Bromophenyl)ethylamine 98%
1-(4-Bromophenyl)ethylamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00025548
CAS Number: 24358-62-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR