6-Bromo-2-chloro-3-ethylquinoline 98+%

Supplier: Ambeed
Danger

A399155-100MG A399155-1G A399155-250MG
77033-626EA 43 USD
77033-626 77033-628 77033-630
6-Bromo-2-chloro-3-ethylquinoline 98+%
6-Bromo-2-chloro-3-ethylquinoline

Precaución: for Research Use Only

Formula: C₁₁H₉BrClN
MDL Number: MFCD09029946
CAS Number: 409346-70-9
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR