2-Thiouracil 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxypyrimidine-2(1H)-thione

A399147-1000G A399147-100G A399147-1MG A399147-25G A399147-500G
77003-870EA 140.04 USD
77003-870 77003-872 77003-874 77003-876 77003-878
2-Thiouracil 97%
2-Thiouracil

Precaución: for Research Use Only

Formula: C₄H₄N₂OS
MW: 128.15 g/mol
MDL Number: MFCD00006039
CAS Number: 141-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR