6-Chloro-2-methylindole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-methyl-1H-indole

A399055-1G A399055-250MG A399055-5G
77033-618EA 84 USD
77033-618 77033-620 77033-622
6-Chloro-2-methylindole 98%
6-Chloro-2-methylindole

Precaución: for Research Use Only

Formula: C₉H₈ClN
MW: 165.62 g/mol
MDL Number: MFCD09027716
CAS Number: 6127-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR