Cyclopropyl(pyridin-3-yl)methanamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-1-(3-pyridyl)methylamine

A398861-1G A398861-250MG
76990-204EA 215.37 USD
76990-204 76990-206
Cyclopropyl(pyridin-3-yl)methanamine 95%
Cyclopropyl(pyridin-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₉H₁₂N₂
MW: 148.205 g/mol
MDL Number: MFCD05191947
CAS Number: 535925-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR