3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-(3-hydroxypropyl)pyrazole

A398366-1G A398366-250MG A398366-5G
77003-864EA 49.87 USD
77003-864 77003-866 77003-868
3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol 98%
3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol

Precaución: for Research Use Only

Formula: C₆H₉BrN₂O
MW: 205.06 g/mol
MDL Number: MFCD06804869
CAS Number: 925180-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR