2,6-Di-tert-butylphenol 98%

Supplier: Ambeed
Warning

A397955-1000G A397955-100G A397955-25G A397955-500G
77003-856EA 24 USD
77003-856 77003-858 77003-860 77003-862
2,6-Di-tert-butylphenol 98%
2,6-Di-tert-butylphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O
MDL Number: MFCD00008820
CAS Number: 128-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR