4-Amino-1-phenylpyrrolidin-2-one 95%

Supplier: Ambeed
Warning

A397421-100MG A397421-1G A397421-250MG
77016-270EA 70 USD
77016-270 77016-272 77016-274
4-Amino-1-phenylpyrrolidin-2-one 95%
4-Amino-1-phenylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD11986777
CAS Number: 3026-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR