5-(2-Chlorophenyl)-1,3,4-thiadiazol-2-amine 95%
Supplier: Ambeed
|
Warning
|
A397360-1G
A397360-250MG
A397360-25G
77003-844EA
13.8
USD
77003-844
77003-846
77672-960
5-(2-Chlorophenyl)-1,3,4-thiadiazol-2-amine 95%
5-(2-Chlorophenyl)-1,3,4-thiadiazol-2-amine
Precaución: for Research Use Only
Formula:
C₈H₆ClN₃S MW: 211.67 g/mol |
MDL Number:
MFCD00548482 CAS Number: 828-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...