11-Bromo-1-undecanol 97%

Supplier: Ambeed
Warning

A396874-100G A396874-10G A396874-25G A396874-500G A396874-5G A396874-1000G
77003-826EA 84 USD
77003-826 77003-828 77003-830 77003-832 77003-834 77882-059
11-Bromo-1-undecanol 97%
11-Bromo-1-undecanol

Precaución: for Research Use Only

Formula: HO(CH₂)₁₀CH₂Br
MDL Number: MFCD00004752
CAS Number: 1611-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR