6-Bromo-1,2-benzisoxazol-3-ol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromobenzo[d]isoxazol-3(2H)-one
A396845-100MG
A396845-1G
A396845-250MG
77003-820EA
24.41
USD
77003-820
77003-822
77003-824
6-Bromo-1,2-benzisoxazol-3-ol 97%
6-Bromo-1,2-benzisoxazol-3-ol
Precaución: for Research Use Only
Formula:
C₇H₄BrNO₂ MW: 214.02 g/mol |
MDL Number:
MFCD13193078 CAS Number: 65685-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...