(S)-1-(4-(Methylthio)phenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A396464-1G A396464-250MG
76990-186EA 296 USD
76990-186 76990-188
(S)-1-(4-(Methylthio)phenyl)ethanamine hydrochloride 95%
(S)-1-(4-(Methylthio)phenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNS
MW: 203.73 g/mol
MDL Number: MFCD16295039
CAS Number: 1206910-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR