N-(5-Chloropyrimidin-2-yl)acetamide 95%

Supplier: Ambeed
Warning

A396300-100MG A396300-1G
77016-256EA 14 USD
77016-256 77890-815
N-(5-Chloropyrimidin-2-yl)acetamide 95%
N-(5-Chloropyrimidin-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MDL Number: MFCD24557279
CAS Number: 68303-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR