(+)-Usnic acid 95%

Supplier: Ambeed
Warning

Synonyms: (9bR)-2,6-Diacetyl-7,9-dihydroxy-8,9b-dimethyldibenzo[b,d]furan-1,3(2H,9bH)-dione

A395779-100MG A395779-5G A395779-25G
77033-610EA 5.31 USD
77033-610 77601-748 77672-956
(+)-Usnic acid 95%
(+)-Usnic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₇
MW: 344.32 g/mol
MDL Number: MFCD00016878
CAS Number: 7562-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR