Methyl 3-chloro-4-hydroxyphenylacetate 95%
Supplier: Ambeed
|
Warning
|
A395402-100MG
A395402-1G
A395402-250MG
A395402-25G
A395402-5G
77003-810EA
16.98
USD
77003-810
77003-812
77003-814
77003-816
77003-818
Methyl 3-chloro-4-hydroxyphenylacetate 95%
Methyl 3-chloro-4-hydroxyphenylacetate
Precaución: for Research Use Only
Formula:
C₉H₉ClO₃ MW: 200.62 g/mol |
MDL Number:
MFCD00209663 CAS Number: 57017-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...