1-(2-Tetrahydropyranyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole

A395367-100G A395367-10G A395367-1G A395367-25G A395367-5G
77033-600EA 65.78 USD
77033-600 77033-602 77033-604 77033-606 77033-608
1-(2-Tetrahydropyranyl)-1H-pyrazole 98%
1-(2-Tetrahydropyranyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MW: 152.20 g/mol
MDL Number: MFCD11044692
CAS Number: 449758-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR