3-Aminofluoranthene 96%

Supplier: Ambeed
Warning

Synonyms: Fluoranthen-3-amine

A395252-1G A395252-250MG A395252-5G
77033-592EA 20.16 USD
77033-592 77033-594 77033-596 77033-598
3-Aminofluoranthene 96%
3-Aminofluoranthene

Precaución: for Research Use Only

Formula: C₁₆H₁₁N
MW: 217.2700 g/mol
MDL Number: MFCD00001185
CAS Number: 2693-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR